missing translation for 'onlineSavingsMsg'
Learn More

KLRG1 Mouse anti-Human, APC, Clone: 13F12F2, eBioscience™

Mouse Monoclonal Antibody

Brand:  Affymetrix eBioscience 17-9488-42

Code : NEW

Additional Details : Weight : 0.23750kg

 View more versions of this product

Product Code. 15588246

  • £228.00 / 100 tests

EU Stock 3

Please to purchase this item. Need a web account? Register with us today!


Description

Description

KLRG1 (Killer cell lectin-like receptor G1, MAFA, 2F1-Ag) is a homodimeric member of the lectin-like type 2 transmembrane receptor family whose members contain characteristic immunoreceptor tyrosine-based inhibitory motifs (ITIMs) in their cytoplasmic domains. These ITIMs interact with the SH2 domains of protein phosphatases such as SHP-1. KLRG1 is an inhibitory receptor that is expressed on natural killer (NK) cells and certain T cells. NK cells are involved in the lysis of tumor cells and virus-infected cells, and mediate humoral and cell-mediated immune responses.
Specifications

Specifications

KLRG1
Monoclonal
5 μL/Test
PBS with 0.1% gelatin, 0.2% BSA and 0.09% sodium azide; pH 7.2
Q96E93
2F1, CLEC15A, MAFA, MAFA-2F1, MAFA-L, MAFA-LIKE, C-type lectin domain family 15 member A, C-type lectin domain family 15, member A, ITIM-containing receptor MAFA-L, MAFA-like receptor, killer cell lectin-like receptor subfamily G member 1, mast cell function-associated antigen (ITIM-containing)
Mouse
Affinity chromatography
RUO
Antibody
Monoclonal
Human
Flow Cytometry
13F12F2
APC
KLRG1
Liquid
KLRG1
IgG2a, kappa
100 Tests
4° C, store in dark, DO NOT FREEZE!
Primary
10219
Videos
Documents

For Research Use Only.