missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KLRG1 Golden Hamster, V450, Clone: 2F1, enQuireBio™
Golden Hamster Monoclonal Antibody
Brand: EnquireBio QAB74-V450-100ug
Additional Details : Weight : 0.01000kg
Specifications
KLRG1 | |
Monoclonal | |
0.5mg/mL | |
10 mM NaH2PO4, 150 mM NaCl, 0.09% NaN3, 0.1% gelatin, pH7.2 | |
Klrg1 | |
Protein A/G Purified | |
RUO | |
50928 | |
IgG |
Flow Cytometry | |
2F1 | |
Violet Fluor 450 (V450) | |
NP_058666 | |
Golden Syrian Hamster | |
100 μg | |
Primary | |
Store at 2-4°C. |