missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KLRG1 Golden Hamster, V450, Clone: 2F1, enQuireBio™
Golden Hamster Monoclonal Antibody
£109.00 - £256.00
Specifications
Antigen | KLRG1 |
---|---|
Clone | 2F1 |
Concentration | 0.5mg/mL |
Applications | Flow Cytometry |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15962428
|
EnquireBio
QAB74-V450-25UG |
25 μg |
£109.00
25µg |
Estimated Shipment: 16-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15952428
|
EnquireBio
QAB74-V450-100UG |
100 μg |
£256.00
100µg |
Estimated Shipment: 16-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
KLRG1 | |
0.5mg/mL | |
Monoclonal | |
Golden Syrian Hamster | |
10 mM NaH2PO4, 150 mM NaCl, 0.09% NaN3, 0.1% gelatin, pH7.2 | |
50928 | |
IgG | |
Protein A/G Purified |
2F1 | |
Flow Cytometry | |
Violet Fluor 450 (V450) | |
RUO | |
NP_058666 | |
Klrg1 | |
Primary | |
Store at 2-4°C. |