missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KLHL2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA550436
Additional Details : Weight : 0.01000kg
Description
The antibody detects endogenous levels of total KLHL2 protein.
KLHL2 Polyclonal antibody specifically detects KLHL2 in Human samples. It is validated for Immunohistochemistry (Paraffin), Western Blot.Specifications
KLHL2 | |
Polyclonal | |
Unconjugated | |
KLHL2 | |
6030411N21Rik; ABP-KELCH; actin-binding protein Mayven; AU020744; kelch; kelch like family member 2; kelch-like 2, Mayven; kelch-like family member 2; kelch-like protein 2; KLHL2; MAV; MAYVEN; si:ch211-287c11.1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
11275 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.9 mg/mL | |
PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 | |
O95198 | |
KLHL2 | |
Fusion protein corresponding to a region derived from internal residues of human kelch-like family member 2 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |