Learn More
KLHL14 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA555179
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: IGGNHLKGFS HLDVMLVECY DPKGDQWNIL QTPILEGRSG PGCAVLDDSI YLVGGYSWSM GAYKSSTICY CPEKGTWTEL EGDVAEPLAG PACVTVILPS C Highest antigen sequence identity to the following orthologs: Mouse - 98%, Rat - 98%.
KLHL14 Polyclonal antibody specifically detects KLHL14 in Human samples. It is validated for Immunohistochemistry.Specifications
KLHL14 | |
Polyclonal | |
Unconjugated | |
KLHL14 | |
6330403N15Rik; AI785250; kelch like family member 14; kelch-like 14; kelch-like family member 14; kelch-like protein 14; Kiaa1384; KLHL14; Printor; protein interactor of Torsin-1A; protein interactor of torsinA; RGD1566178; si:dkeyp-7a3.2 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
57565 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9P2G3 | |
KLHL14 | |
Recombinant protein corresponding to Human KLHL14 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only