Learn More
KLHDC8B Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA552566
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: GTVAHQDGHL LVLGGCGRAG LPLDTAETLD MASHTWLALA PLPTARAGAA AVVLGKQVLV VGGVDEVQSP VAAVEAFLMD EGRWERRATL PQAAMGVATV ERDGMVYALG Highest antigen sequence identity to the following orthologs: Mouse - 98%, Rat - 99%.
KLHDC8B Polyclonal antibody specifically detects KLHDC8B in Human samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot.Specifications
KLHDC8B | |
Polyclonal | |
Unconjugated | |
KLHDC8B | |
4931406O17Rik; CHL; FLJ11302; FP17659; kelch domain containing 8B; kelch domain-containing protein 8B; KLHDC8B; MGC35097 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
200942 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q8IXV7 | |
KLHDC8B | |
Recombinant protein corresponding to Human KLHDC8B | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only