missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KLF6 Sheep anti-Human, Polyclonal, R&D Systems™
Sheep Polyclonal Antibody has been used in 1 publication
POA
Specifications
Antigen | KLF6 |
---|---|
Immunogen | E. coli-derived recombinant human KLF6 Met1-Ser109 Accession # Q99612 |
Gene ID (Entrez) | 1316 |
Gene Accession No. | Q99612 |
Formulation | Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative |
Description
KLF6 Polyclonal specifically detects KLF6 in Human samples. It is validated for Western Blot.Specifications
KLF6 | |
1316 | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
Primary | |
Detects human KLF6 in direct ELISAs and Western blots. In direct ELISAs and Western blots, this antibody shows less than 5% cross-reactivity with rhKLF4, rhKLF5 and rmKLF15. | |
Unconjugated | |
Western Blot | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
E. coli-derived recombinant human KLF6 Met1-Ser109 Accession # Q99612 | |
Q99612 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Sheep | |
IgG | |
Affinity Purified | |
RUO |
For Research Use Only