Learn More
Ecolab™ Klerwipe™ 70/30 IPA Blended With WFI
Sterile heavy weight, pre-laundered, ultrasonically sealed edge 100% polyester wipes impregnated with 70% isopropyl alcohol blended with Water for Injection (WFI) with endotoxin level guaranteed to be less than 1 EU/wipe
Brand: Ecolab™ 3079400
Code : NEW
Additional Details : Weight : 2.47500kg Transport : UN number : 3175 Chem class : 4.1 Pack group : II
Description
- The alcohol blend is 0.2 micron filtered then filled and triple bagged in a Grade C cleanroom before being terminally sterilised by gamma irradiation.
- A sterile ready-to-use wipe in a resealable pouch, featuring Klertear on outer wraps, for high grade small surfaces.
Specifications
Pouch Wipe Blended with WFI | |
20 x 10 Wipes |
Safety and Handling
- Danger
Hazard Category
- Flammable liquid Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H226-Flammable liquid and vapour.
- H336-May cause drowsiness or dizziness.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.