Learn More
Ecolab™ Klerwipe™ 70/30 IPA
Sterile low particulate wipes impregnated with 70% isopropyl alcohol blended with deionised water
£636.00
Product Code | Brand | Description | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Description | Price | Quantity & Availability | |||||
15346118
|
Ecolab™
3079390 |
Pouch Wipe in VHP |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
- The alcohol blend is 0.2 micron filtered then filled and double bagged in a Grade C cleanroom before being terminally sterilised by gamma irradiation
- A sterile ready-to-use wipe for small surfaces available in either a resealable pouch featuring Klertear on outer wrap, or in a tub
Specifications
70% Isopropyl Alcohol |