Learn More
Kimberly-Clark™ KLEENGUARD™ G20 Nitrile Gloves
Kimberly-Clark™ KLEENGUARD™ G20 Nitrile Gloves are an ambidextrous glove that provides protection against chemical splash in a wide range of applications.
Brand: Kimberly-Clark™ 90093
Code : NEW
Additional Details : Weight : 0.01000kg
Description
- PPE Category 3 protection
- For increased protection against chemical splashes of several chemicals.
- Provides exceptional touch sensitivity and textured fingertips to improve grip and the handling of small objects
- AQL 0.65 for pinholes guarantees quality of production process, waste reduction and better protection
- Independently accredited
- Suitable for food handling
- Beaded cuff for added strength and ease of donning
- Latex Free
- Powder free minimises srying effects of pwder and skin allergies
- Ambidextrous
- Portable dispenser box for mobility and easy access
Specifications
Atlantic Green | |
Textured | |
Beaded/Rolled | |
G20 | |
Gloves |
PPE Category III | |
Ideal for providing PPE category 3 protection against chemical splash | |
2500Pack | |
Large | |
0.06 mm |