missing translation for 'onlineSavingsMsg'
Learn More

Kimberly-Clark™ Kleenguard* A50 Breathable Splash & Particle Protection Coveralls

Short-life, breathable hooded coveralls with storm flap for respirator. Kimberly-Clark™ KLEENGUARD™ A50 Breathable Splash and Particle Protection Hooded Coveralls, White are specially designed for comfort, safety and strength.

Brand:  Kimberly-Clark™ 96840

Code : 94

Additional Details : Weight : 7.40000kg

 View more versions of this product

Product Code. 11740684

  • £335.00 / Pack of 25


Estimated Shipment
12-02-2018
Add to basket

Description

Description

  • White, limited life hooded coverall
  • Supplied in packs of 25
  • Provides PPE Category III protection
  • Storm flap and hood designed for easier taping to a respirator
  • Provides anti-static and general protection against water-based or chemical splashes, liquids, and dust particles
  • Durable SMS fabric with additional treatment for increased protection
  • Available in sizes Small through 3X-Large
Specifications

Specifications

Two-way zip
Unisex
White
60cm
A50 PPE Category III
SMS
Stitched
X-Large
6.28 kg
A50 Breathable Splash & Particle Protection Coverall
Videos
Documentation