missing translation for 'onlineSavingsMsg'
Learn More

Kimberly-Clark™ KLEENEX™ 27 Toilet Tissue Bulk Pack

Dispensed one at a time so they are touched only by the user. Kimberly-Clark™ KLEENEX™ 27 Toilet Tissue Bulk Pack tissues dissolve in cold water to reduce blocked drains.

Brand:  Kimberly-Clark™ 4477

Code : 02

Additional Details : Weight : 1.00000kg

Product Code. 12734236

  • £42.40 / Pack of 27


Estimated Shipment
07-06-2018
Add to basket

Description

Description

  • Supplied in packs of 27 x 260 white, 2-ply folded toilet tissue sheets
  • Embossed with the Kleenex™ brand
  • Dissolve in water, preventing blockages and reducing maintenance
  • Manufactured from 100% recycled, FSC-certified paper paper
  • Dispensed one at a time so they are touched only by the user
  • Help reduce costs and waste with up to 40% less used compared to rolls
  • Bulk packaging helps to reduce contamination and the spread of germs
  • ISO 14001, ISO 9001, EU Ecolabel and FSC certified
  • Ideal for office and hotel washrooms that have a high-quality image
  • For use with the wall-mounted AQUARIUS™ Toilet Tissue Dispenser
Specifications

Specifications

ISO 14001, ISO 9001, EU Ecolabel, FSC certified
18.6 x 12.5cm
Paper
2-Ply
Ideal for office and hotel washrooms that have a high-quality image
Toilet Tissue
White
Folded
27 x 260 xheets
12.5cm
18.6cm
Videos