Learn More
Invitrogen™ KIT [Y823D] Recombinant Human Protein
KIT (c-KIT) Y823D encodes the ligand-binding chain (alpha) of the gamma interferon receptor which is a heterodimer of IFNGR1 and IFNGR2. Recombinant human KIT Y823D (544-end) was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag.
£431.00 - £6530.00
Specifications
For Use With (Application) | Enzyme and Protein Activity Assay, Industrial and Applied Science, Kinase Assay, Kinase Biology, Kinase Proteins and Expression Reagents, Pharma and Biopharma, Protein Assays and Analysis, Protein Biology, Target and Lead Identification and Validation |
---|---|
Formulation | 50mM Tris-HCl pH 7.5, 150mM NaCl, 0.25mM DTT, 10mM glutathione, 0.1mM EDTA, 0.1mM PMSF, 25% glycerol |
Molecular Weight (g/mol) | 75.2kDa |
Storage Requirements | Store at -68 to -85°C. Aliquot at first use to avoid multiple freeze-thaws. |
Gene Alias | PBT, SCFR, CD119 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15448963
|
Invitrogen™
PV6322 |
10μg |
£431.00
10µg |
Estimated Shipment: 03-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15458963
|
Invitrogen™
PV6323 |
100μg |
£1460.00
100µg |
Estimated Shipment: 13-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15468963
|
Invitrogen™
PV6324 |
1mg |
£6530.00
1mg |
Estimated Shipment: 13-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
KIT Y823D is a regulator of hematopoietic stem cells during homeostasis and under conditions of infectious stress. A genetic variation in KIT Y823D is associated with susceptibility to Helicobacter pylori infection. In addition, defects in KIT Y823D are a cause of mendelian susceptibility to mycobacterial disease, also known as familial disseminated atypical mycobacterial infection. KIT Y823D is also associated with the clinical outcome of HBV infection in Chinese adults.
- Validated in LanthaScreen™ Kinase Binding Assay
- Protocols available for simple use and optimized performance
- Multiple pack sizes available for convenient use
Specifications
Enzyme and Protein Activity Assay, Industrial and Applied Science, Kinase Assay, Kinase Biology, Kinase Proteins and Expression Reagents, Pharma and Biopharma, Protein Assays and Analysis, Protein Biology, Target and Lead Identification and Validation | |
75.2kDa | |
PBT, SCFR, CD119 | |
Human | |
GST Tag |
50mM Tris-HCl pH 7.5, 150mM NaCl, 0.25mM DTT, 10mM glutathione, 0.1mM EDTA, 0.1mM PMSF, 25% glycerol | |
Store at -68 to -85°C. Aliquot at first use to avoid multiple freeze-thaws. | |
aa 544-end | |
Recombinant |