missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ KIT Ligand, human

£68.71 - £128.00

Specifications

Molecular Weight (g/mol) 18.7 kd
Packaging Plastic bottle
Content And Storage Store at -20°C
Form Lyophilized
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15458629
SDS View Documents Promotion Details
Thermo Scientific™
J67079.!E
10μg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
15468629
SDS View Documents Promotion Details
Thermo Scientific™
J67079.&E
2μg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Specifications

18.7 kd
Plastic bottle
Store at -20°C
Lyophilized
Videos
SDS
Documents

Documents

Product Certifications