Learn More
Thermo Scientific™ KingFisher Apex Benchtop Sample Prep
KingFisher Apex is the most user-friendly and versatile benchtop sample prep instrument in the lab for 24 and 96 samples per run. Consistent high-throughput extraction of DNA, RNA, proteins and cells with extensive selection of validated application protocols
£51000.00 - £51100.00
Products 4
Product Code Brand Description Price Quantity & Availability  
Product Code Brand Description Price Quantity & Availability  
16592442
View accessory products View Documents
Thermo Scientific™
5400910
KingFisher Apex with 96 PCR head
£51100.00
Each
View alternative products
Save Up to 
»
Estimated Shipment
29-04-2021
16502452
View accessory products View Documents
Thermo Scientific™
5400920
KIngFisher Apex with 96 Combi head
£51100.00
Each
View alternative products
Save Up to 
»
Estimated Shipment
29-04-2021
16512452
View accessory products View Documents
Thermo Scientific™
5400930
KingFisher Apex with 96 DW head
£51000.00
Each
View alternative products
Save Up to 
»
Estimated Shipment
29-04-2021
16522452
View accessory products View Documents
Thermo Scientific™
5400940
KingFisher Apex with 24 Combi head
£51100.00
Each
View alternative products
Save Up to 
»
Estimated Shipment
29-04-2021
Description
Description
KingFisher Apex is a versatile and user-friendly instrument which uses magnetic particle processing to automate the purification or isolation of DNA, RNA, proteins, and cells from virtually any source. The Apex moves magnetic beads not liquids through the main steps of sample preparation -- bind, wash, elute. This process provides excellent quality, reproducible results, and reduces hands-on time for sample processing. 
  • Purify 24 or 96 samples in a single run
  • Easy to use interface with touch screen empowering the user to control all details of the application and purification process
  • Free BindIx PC software for added convenience
  • Extensive selection of validated protocols for different type of applications
  • Cloud library for quick protocol retrieval
  • Choose from 24, 96 deep-well, and PCR formats, which includes corresponding magnet heads, tip combs, and plates
SDS