missing translation for 'onlineSavingsMsg'
Learn More

Kinesis™ Omnifit™ Frits for Chromatography Columns

Brand:  Kinesis™ 006FR-15-10

Code : Z3

Additional Details : Weight : 0.20000kg

 View more versions of this product

Product Code. 13191304

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance
View alternative products »


Specifications

Specifications

15mm
Empty Glass chromatography columns
10μm
15mm
PTFE
Universal frit
Videos