Learn More
KINEMATICA Polytron™ PT1200E Handheld Homogenizer
Brand:  KINEMATICA 11010028
Code : 04
Additional Details : Weight : 1.00000kg
Product Code. 13160633
  • £1100.00 / Each
Call for latest availability
+44 (0)1509 555 500
Add to basket
Specifications
Specifications
Ideal for pulverizing plant and animal tissue, grain and seed, soil extraction, DNA/RNA extraction, Karl Fischer sample prep, toxicology and food science
Drive Unit
PT 1200E
Videos
Documentation View accessory products