missing translation for 'onlineSavingsMsg'
Learn More

KINEMATICA Polytron™ Model PT 10-35 GT / PT 3100 D Homogenizer Accessory, Generator

Powerful and versatile benchtop homogenizer. KINEMATICA Polytron™ Model PT 10-35 GT-D Homogenizer completely homogenizes samples from 0.1 to 4000mL.

Brand:  KINEMATICA 11030026

Code : 04

Additional Details : Weight : 0.50000kg

 View more versions of this product

Product Code. 10535613

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance


Description

Description

  • Completely homogenizes samples from 0.1 to 4000mL
  • Powerful 1200W motor provides quiet and reproducible results
  • Homogenize small tissues to fine emulsions and suspensions
  • Get the right results with a variety of generators
Specifications

Specifications

Saw Tooth
PT 10-35 GT and PT 3100 D
0.3 to 10mL
PT-DA 07/2EC-B101
Videos
Documentation