Learn More
Alfa Aesar™ Kinase Storage Buffer
For cellular and molecular biology applications
Brand:  Alfa Aesar™ J63719.AE
Code : NEW
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 15455909
  • £27.98 / 100mL
Estimated Shipment date
from Supplier 07-03-2018
Add to basket
Disclaimers:
For Research Use Only
Description
Description
  • Formulation: 10mM Tris, 50mM sodium chloride, 1mM magnesium chloride, and 50% glycerol, pH 7.5
Specifications
Specifications
Kinase Storage Buffer
Liquid
10mM Tris, 50mM sodium chloride, 1mM magnesium chloride, and 50% glycerol, pH 7.5
100mL