missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Kinase buffer V (5X), Thermo Scientific™
For cellular and molecular biology applications
£34.15 - £42.19
Specifications
Components | 100mM MOPS, 50mM magnesium chloride, and 50mM manganese chloride, pH 7.4 |
---|---|
Content And Storage | Ambient temperatures |
Product Type | Kinase buffer V (5X) |
Form | Liquid |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15494499
|
Thermo Scientific Alfa Aesar
J62045.AK |
250 mL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
15484499
|
Thermo Scientific Alfa Aesar
J62045.AE |
100 mL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
- Formulation:100mM MOPS, 50mM magnesium chloride, and 50mM manganese chloride, pH 7.4
Specifications
100mM MOPS, 50mM magnesium chloride, and 50mM manganese chloride, pH 7.4 | |
Ambient temperatures | |
Kinase buffer V (5X) | |
Liquid |
For Research Use Only