Learn More
Kimberly-Clark™ Professional Kimtech Science™ Comfort Nitrile Gloves
Experience the feeling of high comfort, combined with quality. Kimtech Science™ Comfort Nitrile Gloves are specifically engineered for protection in low risk research applications.
Brand:  Kimberly-Clark™ Professional 47676
Code : NEW
Additional Details : Weight : 0.80000kg
 View more versions of this product
Product Code. 15362206
  • £9.95 / Pack of 150
In Stock 52
Add to basket
Description
Description
  • Combines high comfort, quality and cleanliness for protection in low risk research applications
  • A brand-new nitrile formulation engineered for comfort
  • 150 gloves per standard sized box for all sizes
  • Better chemical protection than latex, tested on the 6 most common chemicals in the lab
  • EPI Category III
Specifications
Specifications
Nitrile
150/Box
White
X-Large
Powder-free
Videos
Documentation