Learn More
Kimberly-Clark™ Kimtech Pure™ A6 Clean Overalls
Maintain ISO 7 Class C or higher cleanroom environments with breathable, ultra light overalls. The Kimtech Pure™ A6 Clean Overalls are made of a high performance, chemical resistant fabric that are origami folded to reduce gowning time.
Brand:  Kimberly-Clark™ 47686
Code : NEW
Additional Details : Weight : 5.38000kg
 View more versions of this product
Product Code. 15518881
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance
View alternative products »
Description
Description
  • High performance chemical resistant fabric
  • Origami folded to reduce gowning time
  • Presented unzipped and inside-out reducing the risk of contaminating the outside of the garment
Specifications
Specifications
PPE Cat III Type 5/6 (Protection against particles/limited splash protection). Add Infectious agent protection. EN1149-1:1995 Antistatic garment
Collared
Breathable
White
Unhooded
Laminated fabric
High neck
Thumb loops
3X-Large
Videos