Learn More
Kimberly-Clark™ Kimtech Pure™ A6 Clean Overalls
Maintain ISO 7 Class C or higher cleanroom environments with breathable, ultra light overalls. The Kimtech Pure™ A6 Clean Overalls are made of a high performance, chemical resistant fabric that are origami folded to reduce gowning time.
Brand:  Kimberly-Clark™ 47685
Code : NEW
Additional Details : Weight : 6.66000kg
 View more versions of this product
Product Code. 15508881
  • £178.00 / Pack of 25
Call for latest availability
+44 (0)1509 555 500
Add to basket
Description
Description
  • High performance chemical resistant fabric
  • Origami folded to reduce gowning time
  • Presented unzipped and inside-out reducing the risk of contaminating the outside of the garment
Specifications
Specifications
PPE Cat III Type 5/6 (Protection against particles/limited splash protection). Add Infectious agent protection. EN1149-1:1995 Antistatic garment
Collared
Breathable
White
Unhooded
Laminated fabric
High neck
Thumb loops
2X-Large
Videos