Learn More
Kimberly-Clark™ Kimtech Pure™ A6 Clean Overalls
Maintain ISO 7 Class C or higher cleanroom environments with breathable, ultra light overalls. The Kimtech Pure™ A6 Clean Overalls are made of a high performance, chemical resistant fabric that are origami folded to reduce gowning time.
Brand:  Kimberly-Clark™ 47682
Code : NEW
Additional Details : Weight : 5.83000kg
 View more versions of this product
Product Code. 15578871
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
  • High performance chemical resistant fabric
  • Origami folded to reduce gowning time
  • Presented unzipped and inside-out reducing the risk of contaminating the outside of the garment
Specifications
Specifications
Laminated SMS
PPE Cat III Type 5/6 (Protection against particles/limited splash protection). Add Infectious agent protection. EN1149-1:1995 Antistatic garment
White
Collared
Breathable
Standing
Thumb loops
Medium