Learn More
Kimberly-Clark Professional™ Kimtech™ G5 Latex Ambidextrous Gloves
Kimtech™ G5 Latex cleanroom gloves provide sensitive and dextrous protection in ISO Class 5 or higher cleanrooms. The gloves have excellent cleanliness and feature a textured finish for enhanced tactile performance.
Brand:  Kimberly-Clark Professional™ HC4411
Code : NEW
Additional Details : Weight : 12.50000kg
 View more versions of this product
Product Code. 10007321
  • £218.00 / Pack of 1000
Estimated Shipment: 31-08-2022
to see stock.
Description
Description
  • High levels of contamination protection against micro-organisms, chemical splash, particles and extractables
  • Made from non-sterile and silicone-free natural rubber latex
  • Excellent tactile sensitivity and textured finish for handling in delicate processes, in both wet and dry conditions
  • Ambidextrous, powder-free, disposable and featuring a beaded cuff for added strength and easier donning
  • Efficient construction and cleanroom-ready packaging that minimises waste
Specifications
Specifications
ISO Class 5, ISO16604 , IEST-RP-CC005 EN374-1:2003, EN374-3:2003, AQL 1.5 / EN374-2 Level 2, ISO16604
Yes
Yes
Latex
Latex Gloves
Beaded/Rolled
30 cm
White
Exam
Yes
Powdered
Large
1000/Pk.