Learn More
DWK Life Sciences Kimble™ KIMAX™ Brand Filtering Flasks
Designed to afford maximum strength
Brand:  DWK Life Sciences 270504000
Code : NEW
Additional Details : Weight : 1.00000kg
 View more versions of this product
Product Code. 15332324
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Description
Description
  • Heavy walled
  • Necks tooled for uniform stopper fit with a thick flat flange
  • Without side tabulation
  • Durable, white ceramic enamel scale to indicate approximate volume
Certifications
ASTM Specification E1406, Type I requirements
Specifications
Specifications
ASTM, E1406, Type I
4 L
150 to 500 mL
14.96 in.
20.8 cm
8.18 in.
Borosilicate Glass
250 mL
38 cm
12