missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DWK Life Sciences Kimble™ Kontes™ Stainless-Steel Solvent Inlet Filter
Inlet filter for 0.125 in. O.D. Tubing
Brand: DWK Life Sciences 953916-0000
Code : 17
Additional Details : Weight : 1.00000kg
Description
Kontes™ Stainless-Steel Solvent Inlet Filter
Specifications
CTFE nut | |
Inlet Filter | |
0.125 in. O.D. Tubing |
2 µm | |
Stainless Steel |