missing translation for 'onlineSavingsMsg'
Learn More

DWK Life Sciences Kimble™ Kontes™ Ultra-Ware Solvent Inlet Filter, Inlet/Sparge Filter

Ideal for Virtually All Mobile Phases

Brand:  DWK Life Sciences 9539163002

Code : 17

Additional Details : Weight : 1.00000kg

Product Code. 12793666

  • £85.95 / Each


Call for latest availability
+44 (0)1509 555 500
Add to basket

Description

Description

  • 2μm All PEEK Inlet/Sparge Filter
  • OD 1.1 in, Length 0.8 in.
  • Fits Tubing OD 1/8 in.
Specifications

Specifications

PTFE
1 Cs.
Inlet/Sparge Filter
HPLC cap system
Videos