Learn More
DWK Life Sciences Kimble™ Kontes™ Replacement Pestle For All-Glass Dounce Tissue Grinders
Small and large clearance pestles for 15mL and 40mL Dounce Tissue Grinder.
POA
Description
- Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements
Specifications
Dounce all Glass tissue grinders |