Learn More
DWK Life Sciences Kimble™ Kontes™ Adsorption/Ion-Exchange Columns
With or without fritted disc
POA
Specifications
Type | Adsorption/Ion Exchange Chromatography Column |
---|---|
For Use With (Application) | Many separations in adsorption and ion exchange chromatography |
Material | Borosilicate Glass |
Description
- Borosilicate glass
- Sealed-in coarse porosity (40 to 60μm) fritted disc supports packing in K420320 Series columns
- Use glass wool to support packing in K420300 Series columns (packing not included)
Specifications
Adsorption/Ion Exchange Chromatography Column | |
Many separations in adsorption and ion exchange chromatography | |
Borosilicate Glass |