missing translation for 'onlineSavingsMsg'
Learn More

DWK Life Sciences Kimble™ Screw-Thread N-51A Borosilicate Glass Sample Vials: Without Caps

Low extractable Kimax N-51A glass

Brand:  DWK Life Sciences 60910-8

Code : 02

Additional Details : Weight : 0.50000kg

 View more versions of this product

Product Code. 10169672

Quantity Price
1 £688.00 / Pack of 864
5 £650.02 / Pack of 864
Estimated Shipment: 14-03-2022

Add to basket


Description

Description

Low extractable Kimax N-51A glass

  • Borosilicate glass sample vials without caps

Compliance

Kimax N-51A glass conforms to ASTM Type 1, Class B requirements.

Specifications

Specifications

B
25 mm
30 mL
Screw
Screw Cap
ASTM Type I
Clear
Borosilicate Glass
For general purpose
8 dr.
95 mm
Videos
SDS
Documents
Certificates