missing translation for 'onlineSavingsMsg'
Learn More

Kimble™ Aluminum Seals for Headspace Vials

Allow for easy access to vial contents

Brand:  Kimble™ 7382120

Code : 17

Additional Details : Weight : 1.00000kg

 View more versions of this product

Product Code. 10540183

  • £89.80 / Pack of 1000


In Stock 4
Add to basket

Description

Description

  • One piece
  • Fits GPI Aluminum Seal Finish 20/20A
  • Unlined
  • Aluminum
  • 20mm (0.787 in.) size

Model 73820-20

  • Tear-out style
  • Model 73821-20
    • Tear-off style

    Specifications

    Specifications

    Tear-off
    Natural
    One Piece Seal
    20mm
    Aluminum
    1000 Case
    Aluminum
    Videos