missing translation for 'onlineSavingsMsg'
Learn More

Kimberly-Clark™ Professional WypAll™ X90 Cloth Wipers

Brand:  Kimberly-Clark™ Professional 12890

Code : 02

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 12625596

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance
View alternative products »


Specifications

Specifications

White
2-Ply
Ideal for heavy wiping, prepping surfaces with solvents and cleaning Metal shaving
Hydroknit™
16.8 x 8.3 in. (42.67 x 21 cm)
21 cm
42.67 cm
Wypall™ X90 wipes
Videos