Kimberly-Clark™ Professional KleenGuard™ A10 Light-Duty Lab Coat

Lightweight, breathable fabric keep workers cool and comfortable

Overview
Brand: Kimberly-Clark™ Professional

Manufacturer Part Number: 40106

UNSPSC: 46181532

Code: 94

Additional Details:
Additional Details: Weight: 0.01000kg



Product Code. 12026659

Quantity Price
1 £ 287.0 / Pack of 50
Estimated Shipment
06-04-2017
Add to basket


Description and Specification

Specification

Material Cotton
Material Polypropylene
Closure Type Snap
Disposable Yes
Size 3X-Large
Cuff Style Elastic
Color White
For Use With (Application) Suited for food processing, general maintenance, manufacturing
Certifications/Compliance ANSI ISEA 101-1996