missing translation for 'onlineSavingsMsg'
Learn More

Kimberly-Clark™ Professional Kimtech Pure™ G5 Latex Gloves

Manufacturer-suggested ISO Class 5 or higher

Brand:  Kimberly-Clark™ Professional HC2211

Code : 03

Additional Details : Weight : 9.10000kg

 View more versions of this product

Product Code. 11740783

  • £178.00 / Pack of 100


In Stock 5
Add to basket

Alert:

These products contain natural rubber latex, a component of which may cause allergic reaction in some users. Use only as directed and in accordance with manufacturer instructions and warnings. Please consult your institution's policies regarding use of these products.


Description

Description

Low in natural rubber proteins.

  • Textured palm and fingers
  • Beaded cuff adds strength and makes donning easier
  • Ambidextrous
  • Thickness: 8.3mil
  • Length: 10 or 12 in. (25 or 30.5cm)

Order Info

Bulk packed 100 gloves per double-bagged polybag with case liner.

Compliance

Meet or exceed applicable ASTM™ D3578 standards.

Specifications

Specifications

Beaded/Rolled
Rubber
Powder-free
30.5cm
Ambidextrous
Small
Natural
Videos