Kimberly-Clark™ Professional Kimtech Pure™ G5 Latex Gloves

Manufacturer-suggested ISO Class 5 or higher

Brand: Kimberly-Clark™ Professional

Manufacturer Part Number: HC4411

X100 Controlled latex glove 8-8,5

UNSPSC: 42132205

Code: 03

Additional Details:
Additional Details: Weight: 11.50000kg

Product Code. 11760783

Quantity Price
1 £185.15 / Pack of 100
Estimated Shipment
Add to basket

Description and Specification


Color Natural
Glove Cuff Style Beaded
Hand Ambidextrous
Length (Metric) 30.5cm
Material Rubber
Powdered No
Size Large
Thickness 8.3mil

Low in natural rubber proteins.

  • Textured palm and fingers
  • Beaded cuff adds strength and makes donning easier
  • Ambidextrous
  • Thickness: 8.3mil
  • Length: 10 or 12 in. (25 or 30.5cm)