Kimberly-Clark™ Professional Kimtech Pure™ G3 and G5 STERLING™ Nitrile Gloves

Combine the protection of nitrile with the sensitivity of latex

Overview
Brand: Kimberly-Clark™ Professional

Manufacturer Part Number: 98187

UNSPSC: 42132203

Code: S3

Additional Details:
Additional Details: Weight: 9.50000kg



Product Code. 11702473

Quantity Price
1 £ 254.0 / Pack of 1500
Estimated Shipment
19-05-2017
Add to basket


Description and Specification

Specification

Classification ISO Class 5
Exterior Finish Textured
Glove Cuff Style Beaded/Rolled
Hand Ambidextrous
Material Nitrile
Size Large
Powdered Powder-free
Color Gray
Sterility Non-sterile
Length (Metric) 30.48cm