Learn More
Kimberly-Clark™ Professional Kimtech Pure™ A5 Sterile Cleanroom Boots
A high-value combination of quality and comfort
Brand:  Kimberly-Clark™ Professional 12920
Code : NEW
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 15134438
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance
View alternative products »
Description
Description
Engineered to protect workers and your bottom line.
  • Abrasion-resistant SMS (Spunbond Meltblown Spunbond) material
  • Filters out many water-based liquids and dry particulates
  • Easy to don, comfortable to wear and efficient to stock
  • Vinyl edge to reduce slipping and liquid penetration
  • Side grasp ties for secure fit
  • No silicone, no latex, no BHT preservative
  • Triple bagged, vacuum packaged, includes sterility indicator
  • White
Applications:
ISO 5-8 Cleanroom Environments: Compounding, Mixing, Filling, Cleaning.
Specifications
Specifications
White
SMS
Yes
X-Large, 2X-Large
Yes
Videos