Learn More
Kimberly-Clark™ Professional Kimtech Pure™ A5 Sterile Boot Covers with Clean-Don Technology
Innovative Clean-Don technology makes donning easier and reduces risk of contamination
Brand:  Kimberly-Clark™ Professional 31697
Code : 03
Additional Details : Weight : 0.84000kg
 View more versions of this product
Product Code. 15128936
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance
Description
Description
Ideal for wetter applications.
  • Side grasp ties for secure fit
  • Vinyl material covers the whole foot
  • Edge vinyl sole reduces slipping and liquid penetration
  • Seam positioned upwards, away from the floor
  • Type 6 PPE Category III for “parts of the body”
  • Sterility assurance level 10-6
  • Vacuum-sealed packaging
Specifications
Specifications
X-Large/2X-Large
Yes
Videos