Learn More
Kimberly-Clark™ Professional Jackson Safety™ Nemesis™ Eyewear
Sleek, sporty style with lightweight construction
Brand:  Kimberly-Clark™ Professional 25685
Code : NEW
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 12621406
  • £54.40 / Pack of 12
Estimated Shipment
15-06-2018
Add to basket
Description
Description
  • Single, wraparound lens provides 99.9% UV protection
  • Soft-touch temples
  • Polarized lenses offer superior protection from excessive glare; ideal for applications requiring extreme clarity and deep color contrast
  • Base curve 8
Specifications
Specifications
Indoor/Outdoor
Meets ANSI Z87.1-2003 standards.
Black
Videos