missing translation for 'onlineSavingsMsg'
Learn More

Kimberly-Clark Professional™ KleenGuard™ G60 Purple Nitrile™ Cut-Resistant Gloves

Catalog Number 11873480
Click to view available options
Size:
7
8
9
10
11
This item is not returnable. View return policy
This item is not returnable. View return policy

Cut-resistant gloves are constructed of comfortable, breathable fabric and feature purple tapered coating on the fingertips for added dexterity.

High level of cut resistance while enhancing dexterity, grip and comfort

  • Dyneema™ fabric construction
  • Purple Nitrile coated fingertips
  • Flexible dotted palm
  • Ambidextrous

TRUSTED_SUSTAINABILITY
Coating Material Nitrile Dots
Color White, Purple
Cut Resistance Rating ANSI A3
Hand Ambidextrous
Material Dyneema™
Product Type Cut-resistant Gloves
Size 7
EN388 Standard EN 388
For Use With (Application) Glass/Metal handling, Paper making and automotive/aircraft assembly
Glove Cuff Style Knit
Model 97430
Series Kleenguard™ G60
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'