missing translation for 'onlineSavingsMsg'
Learn More

Kimberly-Clark™ Professional Jackson Saftey™ G40 Blue Foam Nitrile-Coated Gloves

Provide 25% better productivity than leather

Brand:  Kimberly-Clark™ Professional 40228

Code : NEW

Additional Details : Weight : 0.80000kg

 View more versions of this product

Product Code. 11863480

  • £40.00 / 12 pairs


Estimated Shipment
08-02-2018
Add to basket

Specifications

Specifications

Palm
Foam
auto parts assembly, materials handling, general maintenance and warehousing
Nylon, Spandex™
Blue
Washable
Knit
Nitrile foam
X-Large
24.99cm
Videos