missing translation for 'onlineSavingsMsg'
Learn More

Kimberly-Clark™ Professional Procedure Masks

22% lighter and cooler than average procedure mask

Brand:  Kimberly-Clark™ Professional 47090

Code : 04

Additional Details : Weight : 2.26000kg

 View more versions of this product

Product Code. 12952495

  • £96.20 / Pack of 500


In Stock 14
Add to basket

Specifications

Specifications

CE
≥96% (BFE), ≥97% (PFE)
Pleated
3-ply construction with white hydro-entangled Polyester binding, fully enclosed soft malleable Aluminum nose piece, Polypropylene, Cellulose, Polyurethane facing components
18cm
White
Yes
0.1 (PFE), 3 (BFE)μ
White tubular knitted Polyester, Polyurethane earloops
Videos
Special Offers

Special Offers

  • Big discounts are available on a range of popular products including chemicals, lab essentials, equipment, consumables, life science and safety products.
    : 13013

    2018-03-31

    *Discount will apply to qualifying orders received by Fisher Scientific between 05/02/2018 and 31/03/2018. Promotion code must be presented at the time of order. Discount applies to the list price in effect at the time the order is received. Promotion cannot be combined with other discounts or promotions. Offer void where prohibited, licensed, or restricted by federal, state, provincial, or local laws or regulation or agency/institutional policy. Other restrictions may apply.

    Terms & Conditions