missing translation for 'onlineSavingsMsg'
Learn More

Kimberly-Clark™ Professional Procedure Masks

22% lighter and cooler than average procedure mask

Brand:  Kimberly-Clark™ Professional 47090

Code : NEW

Additional Details : Weight : 1.00000kg

 View more versions of this product

Product Code. 11823510

  • £8.65 / Pack of 50


In Stock 18
Add to basket

Description

Description

  • Three-layer, pleat-style mask
  • Ear loops
  • Inner facing is light and breathable to reduce irritation
  • Outer facing resists fluid strike through

Specifications

Specifications

CE
≥96% (BFE), ≥97% (PFE)
Pleated
3-ply construction with white hydro-entangled Polyester binding, fully enclosed soft malleable Aluminum nose piece, Polypropylene, Cellulose, Polyurethane facing components
18cm
White
Yes
0.1 (PFE), 3 (BFE)μ
White tubular knitted Polyester, Polyurethane earloops
Videos