missing translation for 'onlineSavingsMsg'
Learn More

Kimberly-Clark™ Professional Kimtech Pure™ G3 Nonsterile Latex Ambidextrous Gloves

Flexibility minimizes fatigue

Brand:  Kimberly-Clark™ Professional HC225

Code : NEW

Additional Details : Weight : 0.80000kg

 View more versions of this product

Product Code. 11867521

  • £28.50 / Pack of 100


Estimated Shipment
09-05-2018
Add to basket

Alert:

These products contain natural rubber latex, a component of which may cause allergic reaction in some users. Use only as directed and in accordance with manufacturer instructions and warnings. Please consult your institution's policies regarding use of these products.


Description

Description

  • Textured palms and fingertips
  • Beaded cuff
  • Thickness: 8.7mil at fingertip
  • Length: 12 in. (30.5cm)

Specifications

Specifications

Beaded/Rolled
Rubber
Powder-free
Non-sterile
Ambidextrous
Small
Natural
30.5cm
Videos