Learn More
KIAA1731NL Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA554842
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: TPRGKKMADP EMLPAGEPRS PAEEEAQQAA SKTDLKTFMG KAQNQKYQGT VKPTFRNGSQ TLSPEAGIFI NKEDSLLYST ESGQETPKLG TLA Highest antigen sequence identity to the following orthologs: Mouse - 34%, Rat - 27%.
KIAA1731NL Polyclonal antibody specifically detects KIAA1731NL in Human samples. It is validated for Immunohistochemistry.Specifications
KIAA1731NL | |
Polyclonal | |
Unconjugated | |
CEP295NL | |
CEP295 N-terminal like; CEP295 N-terminal-like protein; CEP295NL; DDC8; differential display clone 8; KIAA1731 N-terminal like protein; KIAA1731NL; protein DDC8 homolog | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
100653515 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q96MC4 | |
CEP295NL | |
Recombinant protein corresponding to Human KIAA1731NL | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only