missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ Ki 8751
Potent, selective VEGFR-2 inhibitor
POA
Specifications
Chemical Name or Material | N-(2,4-Difluorophenyl)-N'-[4-[(6,7-dimethoxy-4-quinolinyl)oxy]-2-fluorophenyl]urea |
---|---|
CAS | 228559-41-9 |
Assay Percent Range | >97% |
Solubility | Soluble to 100mM in DMSO |
Molecular Formula | C24H18F3N3O4 |
Description
Potent, selective inhibitor of VEGFR-2 tyrosine kinase (IC50 = 0.9 nM). Displays some inhibitory activity towards c-Kit, PDGFRα and FGFR-2 (IC50 values range from 40 to 170 nM) but is highly selective over other receptor tyrosine kinases (IC50 > 10000 nM for FGFR-2, EGFR and HGFR). Inhibits VEGF-stimulated proliferation of human umbilical vein endothelial cells (HUVEC) and inhibits tumor growth in vivo; antiangiogenic.Specifications
N-(2,4-Difluorophenyl)-N'-[4-[(6,7-dimethoxy-4-quinolinyl)oxy]-2-fluorophenyl]urea | |
>97% | |
C24H18F3N3O4 |
228559-41-9 | |
Soluble to 100mM in DMSO | |
Observed MW: 469.41 |
For Research Use Only