missing translation for 'onlineSavingsMsg'
Learn More

Ki-67, PE-eFluor 610, clone: SolA15, eBioscience™

Rat Monoclonal Antibody

Brand:  Affymetrix eBioscience 61-5698-80

Code : NEW

Additional Details : Weight : 0.09500kg

Product Code. 15570517

  • £93.75 / 25µg

Estimated Shipment
28-10-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Description: The monoclonal antibody SolA15 recognizes mouse and rat Ki-67, a 300kDa nuclear protein. Ki-67 is present during all active phases of the cell cycle (G1, S, G2, and mitosis), but is absent from resting cells (G0). Ki-67 is detected within the nucleus during interphase but redistributes to the chromosomes during mitosis. Ki-67 is used as a marker for determining the growth fraction of a given population of cells. In studies of tumor cells, the Ki-67 labeling index refers to the number of Ki-67 positive cells within the population and this is used to predict outcome of particular cancer types. Ki-67 has been shown to interact with the DNA-bound protein chromobox protein homolog 3 (CBX3) (heterochromatin).The SolA15 antibody also recognizes human, non-human primate and canine Ki-67.Applications Reported: This SolA15 antibody has been reported for use in intracellular staining followed by flow cytometric analysis.Applications Tested: This SolA15 antibody has been tested by intracellular staining and flow cytometric analysis of mouse splenocytes using the Foxp3/Transcription Factor Staining Buffer Set (cat. 00-5523) and protocol. Refer to Best Protocols for Staining Protocol (refer to Protocol B: One-step protocol for intracellular (nuclear) proteins). This can be used at less than or equal to 0.125 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.PE-eFluor™ 610 can be excited with laser lines from 488-561 nm and emits at 607 nm. We recommend using a 610/20 band pass filter (equivalent to PE-Texas Red™). Please make sure that your instrument is capable of detecting this fluorochome.Light sensitivity: This tandem dye is sensitive to photo-induced oxidation. Please protect this vial and stained samples from light.Fixation: Samples can be stored in IC Fixation Buffer (cat. 00-8222) (100 μL of cell sample + 100 μL of IC Fixation Buffer) or 1-step Fix/Lyse Solution (cat. 00-5333) for up to 3 days in the dark at 4°C with minimal impact on brightness and FRET efficiency/compensation. Some generalizations regarding fluorophore performance after fixation can be made, but clone specific performance should be determined empirically.Excitation: 488-561 nm; Emission: 607 nm; Laser: Blue Laser, Green Laser, Yellow-Green Laser.Filtration: 0.2μm post-manufacturing filtered.
Specifications

Specifications

Ki-67
Monoclonal
0.2mg/mL
PBS with 0.09% sodium azide; pH 7.2
P46013, E9PVX6
antigen identified by monoclonal antibody Ki-67, antigen KI-67, KIA, proliferation-related Ki-67 antigen, protein phosphatase 1, regulatory subunit 105, RP11-380J17.2
Rat
Affinity chromatography
RUO
Antibody
Monoclonal
Canine, Cynomolgus Monkey, Human, Murine, Non-human Primate, Rat
Flow Cytometry
SolA15
PE-eFluor 610
MKI67
Liquid
MKI67
IgG2a, kappa, kappa
25μg
Store at 2-8°C. Do not freeze. Light-sensitive material. This tandem dye is sensitive to photo-induced oxidation. Protect this vial from light during storage, handling and experimental procedures.
Primary
100686578, 102135895, 17345, 291234, 4288
Videos
Documents

For Research Use Only.