missing translation for 'onlineSavingsMsg'
Learn More

Ki-67, Biotin, clone: SolA15, eBioscience™

Rat Monoclonal Antibody

Brand:  Affymetrix eBioscience 13-5698-82

Code : NEW

Additional Details : Weight : 0.09500kg

 View more versions of this product

Product Code. 15517346

  • £183.54 / 100µg


EU Stock 2
Add to basket

Description

Description

Description: The monoclonal antibody SolA15 recognizes mouse and rat Ki-67, a 300kDa nuclear protein. Ki-67 is present during all active phases of the cell cycle (G1, S, G2, and mitosis), but is absent from resting cells (G0). Ki-67 is detected within the nucleus during interphase but redistributes to the chromosomes during mitosis. Ki-67 is used as a marker for determining the growth fraction of a given population of cells. In studies of tumor cells, the Ki-67 labeling index refers to the number of Ki-67 positive cells within the population and this is used to predict outcome of particular cancer types. Ki-67 has been shown to interact with the DNA-bound protein chromobox protein homolog 3 (CBX3) (heterochromatin).The SolA15 antibody also recognizes human, non-human primate and canine Ki-67.Applications Reported: This SolA15 antibody has been reported for use in intracellular staining followed by flow cytometric analysis, immunohistochemical staining of frozen tissue sections, immunohistochemical staining of formalin-fixed paraffin embedded tissue sections, microscopy, and immunocytochemistry.Applications Tested: This SolA15 antibody has been tested immunocytochemistry of fixed and permeabilized C2C12 cells and can be used at less than or equal to 5 μg/mL or intracellular staining and flow cytometric analysis of stimulated mous esplenocytes cells using the Foxp3/Transcription Factor Buffer Set (cat. 00-5523) and protocol. Please see Best Protocols Section (Staining intracellular Antigens for Flow Cytometry) for staining protocol (refer to Protocol B: One-step protocol for intracellular (nuclear) proteins). This can be used at less than or equal to 0.125 μg per test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.Filtration: 0.2μm post-manufacturing filtered.
Specifications

Specifications

Ki-67
Monoclonal
0.5mg/mL
PBS with 0.1% gelatin and 0.09% sodium azide; pH 7.2
E9PVX6, P46013
antigen identified by monoclonal antibody Ki-67, antigen KI-67, KIA, proliferation-related Ki-67 antigen, protein phosphatase 1, regulatory subunit 105, RP11-380J17.2
Rat
Affinity chromatography
RUO
Antibody
Monoclonal
CanineþCynomolgusþHumanþMurineþNon-human PrimateþRat
Flow CytometryþImmunocytochemistryþImmunohistochemistryþImmunohistochemistry (Frozen)þMicroscopy
SolA15
Biotin
MKI67
Conjugated
Mki67
IgG2a, kappa
100μg
Store at 2-8°C. Do not freeze.
Primary
4288
Videos

For Research Use Only.