missing translation for 'onlineSavingsMsg'
Learn More

Ki-67, eFluor 506, clone: 20Raj1, eBioscience™

Mouse Monoclonal Antibody

Brand:  Affymetrix eBioscience 69-5699-42

Code : NEW

Additional Details : Weight : 0.23750kg

 View more versions of this product

Product Code. 15580687

  • £177.00 / 100 tests

Estimated Shipment date
from Supplier 03-03-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Description: The monoclonal antibody 20Raj1 recognizes the human Ki-67 protein. Two isoforms of Ki-67 exist, a 345 and 395kDa form that are expressed in dividing cells. Ki-67 is expressed in all cell types and is detectable during active phases of the cell cycle (G1, S, G2, and mitosis) but is absent from resting cells (G0). During interphase, Ki-67 expression is localized to the nucleus but redistributes to the chromosomes during mitosis and has specifically been found to associate with heterochromatin-bound proteins such as chromobox protein homolog 3 (CBX3). In studies of tumor cells, Ki-67 expression has been used as a marker for determining the fraction of proliferating cells within a given population of tumor cells.This monoclonal antibody 20Raj1 recognizes canine Ki-67.Applications Reported: This 20Raj1 antibody has been reported for use in intracellular staining followed by flow cytometric analysis.Applications Tested: This 20Raj1 antibody has been pre-titrated and tested by intracellular staining and flow cytometric analysis of stimulated normal human peripheral blood cells using the Foxp3/Transcription Factor Staining Buffer Set (cat. 00-5523) and protocol. Please refer to Best Protocols: Protocol B: One step protocol for (nuclear) intracellular proteins located under the Resources Tab online. This can be used at 5 μL (0.03 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. eFluor™ 506 can be excited with the violet laser line (405 nm) and emits at 506 nm. We recommend using a 510/20 band pass filter, or equivalent. Please make sure that your instrument is capable of detecting this fluorochrome.Excitation: 405 nm; Emission: 506 nm; Laser: Violet Laser.Filtration: 0.2μm post-manufacturing filtered.
Specifications

Specifications

Ki-67
Monoclonal
5μl/Test
PBS with 0.1% gelatin, 0.2% BSA and 0.09% sodium azide; pH 7.2
P46013
antigen identified by monoclonal antibody Ki-67, antigen KI-67, KIA, proliferation-related Ki-67 antigen, protein phosphatase 1, regulatory subunit 105, RP11-380J17.2
Murine
Affinity chromatography
RUO
Antibody
Monoclonal
Canine, Human
Flow Cytometry
20Raj1
eFluor 506
MKI67
Liquid
MKI67
IgG1, kappa, kappa
100 Tests
Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary
100686578, 4288
Videos
Documents

For Research Use Only.